DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals
DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals
DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals
DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals
CAS: 828-01-3 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number ACR427770100
Price (MYR)
1,455.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,455.00
Add to cart
Specifications
Chemical Name or MaterialDL-3-Phenyllactic acid
CAS828-01-3
Melting Point95°C
ColorWhite
EINECS Number212-580-4