N-Acetyl-D-mannosamine (ManNAc), CAS # 7772-94-3, is a monosaccharide sialic acid precursor.
Have Questions?
Change view
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR427530050
Price (MYR)
1,832.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,832.00
Add to cart
Specifications
Chemical Name or MaterialN-Acetyl-D-mannosamine
CAS7772-94-3
CAS Min %97.5
CAS Max %100.0
Health Hazard 2GHS H Statement: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
View more
General Description
N-Acetyl-D-mannosamine (ManNAc) is a precursor used to synthesize N-acetylneuraminic acid, the most common physiologic sialic acid present in humans and other mammals
Sialic acids are terminal sugars on glycoconjugates that can modify biochemistry of glycoproteins and lipids, mediate cellular adhesion, and act as immune regulators
Defects in sialic acid biosynthesis are associated with a number of pathologies
Sialic acids are often found at increased expression levels on tumor cell surfaces
Applications
As a sialic acid precursor, N-Acetyl-D-mannosamine can be used in many glycoengineering applications, such as modifying immune cell activity, imaging glycans, or blocking pathogen binding
N-Acetyl-D-mannosamine hasbeen used in preclinical laboratory studies as a treatment for hereditary inclusion body myopathy, a progressive neuromuscular disease via increased sialylation of podocalyxin
N-Acetyl-D-mannosamine has been used in laboratory studies to induce IgG sialylation and prevent obesity-induced hypertension in mice following high-fat diet
RUO – Research Use Only
General References:
Peng, J.; Vongpatanasin, W.; Sacharidou, A.; Kifer, D.; Yuhanna, I.S.; Banerjee, S.; Tanigaki, K.; Polasek, O.; Chu, H.; Sundgren, N.C.; Rohatgi, A.; Chambliss, K.L.; Lauc, G.; Mineo, C.; Shaul, P.W. Supplementation With the Sialic Acid Precursor N-Acetyl-D-Mannosamine Breaks the Link Between Obesity and Hypertension. Circulation. 2019, 140(24), 2005-2018.
Moons, S.J.; Adema, G.J.; Derks, M.T.; Boltje, T.J.; Büll, C. Sialic acid glycoengineering using N-acetylmannosamine and sialic acid analogs. Glycobiology. 2019, 29(6), 433-445.
Büll, C.; Stoel, M.A.; den Brok, M.H.; Adema, GJ. Sialic acids sweeten a tumor's life. Cancer Res. 2014, 74(12), 3199-204.
Galeano, B.; Klootwijk, R.; Manoli, I.; Sun, M.; Ciccone, C.; Darvish, D.; Starost, M.F.; Zerfas, P.M.; Hoffmann, V.J.; Hoogstraten-Miller, S.; Krasnewich, D.M.; Gahl, W.A.; Huizing, M. Mutation in the key enzyme of sialic acid biosynthesis causes severe glomerular proteinuria and is rescued by N-acetylmannosamine. J Clin Invest. 2007, 117(6), 1585-94.