3,5-Dimethoxybenzyl bromide, 95%
3,5-Dimethoxybenzyl bromide, 95%
3,5-Dimethoxybenzyl bromide, 95%
3,5-Dimethoxybenzyl bromide, 95%
Thermo Scientific Chemicals
3,5-Dimethoxybenzyl bromide, 95%
CAS: 877-88-3 | C9H11BrO2 | 231.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4273400505 gGlass bottle
Catalog number ACR427340050
Price (MYR)
1,015.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,015.00
Add to cart
Chemical Identifiers
CAS4181-05-9
IUPAC Name4-(diphenylamino)benzaldehyde
Molecular FormulaC19H15NO
InChI KeyUESSERYYFWCTBU-UHFFFAOYSA-N
SMILESO=CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow
FormPowder or crystals or crystalline powder
Infrared spectrumConforms
Purity≥97.5%