Specifications
Chemical Name or MaterialTri-n-butylphosphine
CAS998-40-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long la
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
View more