Propionaldehyde, 99+%, AcroSeal™
Propionaldehyde, 99+%, AcroSeal™
Propionaldehyde, 99+%, AcroSeal™
Propionaldehyde, 99+%, AcroSeal™
Propionaldehyde, 99+%, AcroSeal™
Thermo Scientific Chemicals
Propionaldehyde, 99+%, AcroSeal™
CAS: 123-38-6 | C3H6O | 58.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR427211000100 mLAcroSeal™ Glass Bottle
Catalog number ACR427211000
Price (MYR)
511.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
511.00
Add to cart
Specifications
Chemical Name or MaterialPropionaldehyde
CAS123-38-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for