Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
Thermo Scientific Chemicals
Chloroform-d, for NMR, 0.02-0.04 v/v% TMS, 99.8 atom % D, AcroSeal™
CAS: 865-49-6 | CHCl3 | 120.375 g/mol
Have Questions?
Catalog NumberQuantity
ACR426961000100 mL
Catalog number ACR426961000
Price (MYR)
594.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
594.00
Add to cart
Chemical Identifiers
CAS58-85-5
IUPAC Name5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
Molecular FormulaC10H16N2O3S
InChI KeyYBJHBAHKTGYVGT-UHFFFAOYNA-N
SMILESOC(=O)CCCCC1SCC2NC(=O)NC12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with NaOH>=97.5 % (On dried substance)
Specific optical rotation+89° to +93° (20°C, 589 nm) (c=2, 0.1 N NaOH)
View more
For NMR
RUO – Research Use Only