Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™
Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™
Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™
Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™
Thermo Scientific Chemicals
Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™
CAS: 7789-20-0 | H2O | 20.03 g/mol
Have Questions?
Catalog NumberQuantity
ACR426931000100 mL
Catalog number ACR426931000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDeuterium oxide
Density1.1050g/mL
Deuterium (D)99.75% min.
Physical FormLiquid
Refractive Index1.328
View more
For NMR
RUO – Research Use Only