Chloroform-d, for NMR, 99.8 Atom % D, AcroSeal™
Chloroform-d, for NMR, 99.8 Atom % D, AcroSeal™
Chloroform-d, for NMR, 99.8 Atom % D, AcroSeal™
Chloroform-d, for NMR, 99.8 Atom % D, AcroSeal™
Chloroform-d, for NMR, 99.8 Atom % D, AcroSeal™
Thermo Scientific Chemicals
Chloroform-d, for NMR, 99.8 Atom % D, AcroSeal™
CAS: 865-49-6 | CHCl3 | 120.375 g/mol
Have Questions?
Catalog NumberQuantity
ACR426771000100 mL
Catalog number ACR426771000
Price (MYR)
641.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
641.00
Add to cart
Chemical Identifiers
CAS79-41-4
IUPAC Name2-methylprop-2-enoic acid
Molecular FormulaC4H6O2
InChI KeyCERQOIWHTDAKMF-UHFFFAOYSA-N
SMILESCC(=C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.4 %
Color scale=<20 APHA
Appearance (Form)Clear liquid
Refractive index1.4295 to 1.4315 (20°C, 589 nm)
View more
For NMR
RUO – Research Use Only