1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
CAS: 32387-21-6 | C10H9NO2 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR426480010
Price (MYR)
417.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
417.00
Add to cart
Specifications
Chemical Name or Material1-Methylindole-3-carboxylic acid
CAS32387-21-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0