6-Carboxyfluorescein, 96%
6-Carboxyfluorescein, 96%
6-Carboxyfluorescein, 96%
6-Carboxyfluorescein, 96%
Thermo Scientific Chemicals
6-Carboxyfluorescein, 96%
CAS: 3301-79-9 | C21H12O7 | 376.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Catalog number ACR425801000
Price (MYR)
1,474.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,474.00
Add to cart
Chemical Identifiers
CAS16561-29-8
IUPAC Name(1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular FormulaC36H56O8
InChI KeyPHEDXBVPIONUQT-RGYGYFBISA-N
SMILESCCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay from Suppliers CofA≥94.0%
FormPowder, film or foam, solid