Indole-6-carboxylic acid, 98+%, Thermo Scientific™
Indole-6-carboxylic acid, 98+%, Thermo Scientific™
Indole-6-carboxylic acid, 98+%, Thermo Scientific™
Indole-6-carboxylic acid, 98+%, Thermo Scientific™
Thermo Scientific Chemicals
Indole-6-carboxylic acid, 98+%, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR4246800252.5 gGlass bottle
Catalog number ACR424680025
Price (MYR)
621.00
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
621.00
Add to cart
Specifications
Chemical Name or MaterialIndole-6-carboxylic acid
Name Note98+%
CAS1670-82-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: