Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 62-55-5 | C2H5NS | 75.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number ACR424531000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS146982-27-6
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(prop-2-en-1-yloxy)carbonyl]amino}hexanoic acid
Molecular FormulaC25H28N2O6
InChI KeyOJBNDXHENJDCBA-ANBDAQEENA-N
SMILESOC(=O)[C@H](CCCCNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%