Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 62-55-5 | C2H5NS | 75.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR424530250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialThioacetamide
CAS62-55-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause cancer.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water