5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR424515000
Price (MYR)
942.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
942.00
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormWire, 0.25mm (0.01in) diameter, annealed.
Assay from Supplier's CofA≥99.9% (metals basis)
Indicator
RUO – Research Use Only