5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR424511000
Price (MYR)
489.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
489.00
Add to cart
Specifications
Chemical Name or Material5-Sulfosalicylic acid dihydrate
CAS5965-83-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
CAS97-05-2
View more
Indicator
RUO – Research Use Only