L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
CAS: 6106-24-7 | C4H8Na2O8 | 230.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Catalog number ACR424445000
Price (MYR)
1,090.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,090.00
Add to cart
Chemical Identifiers
CAS1450-23-3
SpecificationsSpecification SheetSpecification Sheet
CommentWhite to pale cream
FormPowder
Elemental AnalysisCarbon: 81.30-85.46%
Elemental AnalysisHydrogen: 5.68-5.98%