L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
CAS: 6106-24-7 | C4H8Na2O8 | 230.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number ACR424440025
Price (MYR)
3,114.00
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (MYR)
3,114.00
Add to cart
Specifications
Chemical Name or MaterialL(+)-Tartaric acid, disodium salt, dihydrate
pH7.0 to 9.0 (5% soln. at 25°C)
CAS6106-24-7
ColorWhite
Assay Percent Range99+%