Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number ACR424335000
Price (MYR)
540.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
540.00
Add to cart
Specifications
Chemical Name or MaterialSodium hydroxide
ColorWhite
Boiling Point1390.0°C
CAS1310-73-2
Health Hazard 1Danger