Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Catalog number ACR424330010
Price (MYR)
789.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
789.00
Add to cart
Specifications
Chemical Name or MaterialSodium hydroxide
ColorWhite
Boiling Point1390.0°C
CAS1310-73-2
Health Hazard 1GHS Signal Word: Danger