Acetic acid, sodium salt trihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number ACR424260010
Price (MYR)
572.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
572.00
Add to cart
Specifications
Chemical Name or MaterialAcetic acid, sodium salt trihydrate
pH7.5 to 9.2 (5% soln. at 25°C)
CAS127-09-3
CAS Min %99.0
Health Hazard 1HYGROSCOPIC