Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number ACR424255000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS50-69-1
IUPAC Name(3R,4S,5R)-5-(hydroxymethyl)oxolane-2,3,4-triol
Molecular FormulaC5H10O5
InChI KeyHMFHBZSHGGEWLO-ZHZWXSSZNA-N
SMILESOC[C@H]1OC(O)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<1 % (60°C, 3 h)
Specific optical rotation-18° to -22° (20°C, 589 nm) (c=2, water)
Appearance (Color)White to light beige
Appearance (Form)Powder or crystalline powder