Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
Catalog number ACR424040025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS81029-06-3
IUPAC Namealuminium(3+) nonahydrate triperchlorate
Molecular FormulaAlCl3H18O21
InChI KeyCYRDFMGMNQJULY-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.O.O.O.[Al+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content0.1% max