Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number ACR424032500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS141-43-5
IUPAC Name2-aminoethan-1-ol
Molecular FormulaC2H7NO
InChI KeyHZAXFHJVJLSVMW-UHFFFAOYSA-N
SMILESNCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.4525-1.4565 @ 20?C
Water Content (Karl Fischer Titration)≤1%
View more
Oxidation reagent
RUO – Research Use Only