Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number ACR424030010
Price (MYR)
1,393.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,393.00
Add to cart
Specifications
Chemical Name or MaterialPerchloric acid
Name Noteca. 70% Solution in Water
CAS7601-90-3
Melting Point-18.0°C
Boiling Point203.0°C
View more
Oxidation reagent
RUO – Research Use Only