Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number ACR424005000
Price (MYR)
813.00
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
813.00
Add to cart
Chemical Identifiers
CAS42580-42-7
IUPAC Name2,5-bis(trifluoromethyl)benzoic acid
Molecular FormulaC9H4F6O2
InChI KeyPINBPLCVZSKLTF-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC=C1C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)75.0-81.0?C
Appearance (Color)Pale cream