Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass Bottle
Catalog number ACR424000025
Price (MYR)
1,063.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,063.00
Add to cart
Specifications
Chemical Name or MaterialNitric acid
ColorColorless
CAS7697-37-2
Melting Point-42.0°C
Boiling Point122.0°C