Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number ACR423955000
Price (MYR)
343.00
Add to cart
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
343.00
Add to cart
Chemical Identifiers
CAS367-51-1
IUPAC Namesodium 2-sulfanylacetate
Molecular FormulaC2H3NaO2S
InChI KeyGNBVPFITFYNRCN-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to light pink
Appearance (Form)Powder
Infrared spectrumConforms
Titration Iodimetric>=97.5 %