Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number ACR423950025
Price (MYR)
525.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
525.00
Add to cart
Chemical Identifiers
CAS3425-46-5
IUPAC Namepotassium cyanoselanide
Molecular FormulaCKNSe
InChI KeyKYEKHFSRAXRJBR-UHFFFAOYSA-M
SMILES[K+].[Se-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Titration Argentometric>=98.5 %