Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number ACR423950010
Price (MYR)
445.00
Add to cart
Quantity:
1 L
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
445.00
Add to cart
Chemical Identifiers
CAS95727-86-9
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%