Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Catalog number ACR423835000
Price (MYR)
470.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
470.00
Add to cart
Specifications
Chemical Name or MaterialIsopropanol
CAS67-63-0
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.