Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Catalog number ACR423830025
Price (MYR)
1,012.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,012.00
Add to cart
Chemical Identifiers
CAS16331-48-9
IUPAC Name4-acetamidobenzoyl chloride
Molecular FormulaC9H8ClNO2
InChI KeyKRKXGCJUNKZXOY-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream to yellow to brown
Assay (Titration ex Chloride)≥94.0 to ≤106.0%