Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Catalog number ACR423830010
Price (MYR)
630.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
630.00
Add to cart
Chemical Identifiers
CAS104-90-5
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4950-1.5000 @ 20?C
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to pale yellow