Iodine, Reagent ACS, 99.8%, Thermo Scientific Chemicals
Iodine, Reagent ACS, 99.8%, Thermo Scientific Chemicals
Iodine, Reagent ACS, 99.8%, Thermo Scientific Chemicals
Iodine, Reagent ACS, 99.8%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodine, Reagent ACS, 99.8%, Thermo Scientific Chemicals
CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number ACR423825000
Price (MYR)
1,445.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,445.00
EA
Add to cart
Specifications
Boiling Point185°C
Chemical Name or MaterialIodine
Melting Point113°C
Name Note99.8%
CAS7553-56-2
View more
C-X bond formation reagent
RUO – Research Use Only