Iodic acid, 99.6%, Thermo Scientific Chemicals
Iodic acid, 99.6%, Thermo Scientific Chemicals
Iodic acid, 99.6%, Thermo Scientific Chemicals
Iodic acid, 99.6%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodic acid, 99.6%, Thermo Scientific Chemicals
CAS: 7782-68-5 | ILiO3 | 181.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR423815000
Price (MYR)
3,447.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,447.00
Add to cart
Specifications
Chemical Name or MaterialIodic acid
ColorWhite
Name NoteFor analysis ACS
CAS7782-68-5
Melting Point110.0°C
View more
Oxidation reagent
RUO – Research Use Only