Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
Plastic bottle
Glass Bottle
Catalog number ACR423795001
Price (MYR)
403.00
Add to cart
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
403.00
Add to cart
Chemical Identifiers
CAS99-87-6
IUPAC Name1-methyl-4-(propan-2-yl)benzene
Molecular FormulaC10H14
InChI KeyHFPZCAJZSCWRBC-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4890 to 1.4910 (20°C, 589 nm)
Color scale=<30 APHA
GC>=99.0 %
Specific gravity(25°C) 0.853 to 0.860