Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Plastic bottle
Catalog number ACR423795000
Price (MYR)
427.00
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
427.00
Add to cart
Chemical Identifiers
CAS6294-89-9
IUPAC Namemethoxycarbohydrazide
Molecular FormulaC2H6N2O2
InChI KeyWFJRIDQGVSJLLH-UHFFFAOYSA-N
SMILESCOC(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals or platelets
GC>=96.0 %
Appearance (Color)White to pink
Melting point68°C to 73°C