Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic acid, 47.0 to 49.0%, BrH, CAS Number-10035-10-6, 7732-18-5, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, 126.0 deg.C to 128.0 deg.C, CHEBI:47266
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR423785000
Price (MYR)
741.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
741.00
Add to cart
Chemical Identifiers
CAS107-58-4
IUPAC NameN-tert-butylprop-2-enamide
Molecular FormulaC7H13NO
InChI KeyXFHJDMUEHUHAJW-UHFFFAOYSA-N
SMILESCC(C)(C)NC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder and/or chunks
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
Melting Point126-132?C
C-X bond formation reagent
RUO – Research Use Only