Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic acid, 47.0 to 49.0%, BrH, CAS Number-10035-10-6, 7732-18-5, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, 126.0 deg.C to 128.0 deg.C, CHEBI:47266
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass bottle
Catalog number ACR423780025
Price (MYR)
1,427.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,427.00
Add to cart
Specifications
Chemical Name or MaterialHydrobromic acid
ColorColorless to Yellow
CAS10035-10-6
Melting Point-11.0°C
Boiling Point126.0°C to 128.0°C
View more
C-X bond formation reagent
RUO – Research Use Only