Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
CAS: 10034-93-2 | H4N2·H2SO4 | 130.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
100 g
Packaging:
Plastic bottle
Catalog number ACR423775000
Price (MYR)
1,115.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,115.00
Add to cart
Chemical Identifiers
CAS7637-07-2
IUPAC Nametrifluoroborane
Molecular FormulaBF3
InChI KeyWTEOIRVLGSZEPR-UHFFFAOYSA-N
SMILESFB(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99%
Reduction reagent
RUO – Research Use Only