Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
CAS: 10034-93-2 | H4N2·H2SO4 | 130.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
50 g
500 g
Packaging:
Plastic bottle
Catalog number ACR423771000
Price (MYR)
556.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
556.00
Add to cart
Chemical Identifiers
CAS103-04-8
IUPAC Name2-(phenylsulfanyl)acetate
Molecular FormulaC8H7O2S
InChI KeyMOTOSAGBNXXRRE-UHFFFAOYSA-M
SMILES[O-]C(=O)CSC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Water Content (Karl Fischer Titration)≤1.5%
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥96.0%
View more
Reduction reagent
RUO – Research Use Only