Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
CAS: 10034-93-2 | H4N2·H2SO4 | 130.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR423770500
Price (MYR)
464.00
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
464.00
Add to cart
Chemical Identifiers
CAS620-24-6
IUPAC Name3-(hydroxymethyl)phenol
Molecular FormulaC7H8O2
InChI KeyOKVJCVWFVRATSG-UHFFFAOYSA-N
SMILESOCC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
Melting Point (clear melt)64.0-75.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Reduction reagent
RUO – Research Use Only