Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR423755000
Price (MYR)
635.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
635.00
Add to cart
Chemical Identifiers
CAS502-56-7
IUPAC Namenonan-5-one
Molecular FormulaC9H18O
InChI KeyWSGCRAOTEDLMFQ-UHFFFAOYSA-N
SMILESCCCCC(=O)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4180-1.4220 @ 20?C