Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number ACR423750025
Price (MYR)
1,467.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,467.00
Add to cart
Specifications
Chemical Name or MaterialFormic acid
Name Note96+%
CAS64-18-6
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.