Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur., Thermo Scientific Chemicals
Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur., Thermo Scientific Chemicals
Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur., Thermo Scientific Chemicals
Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur., Thermo Scientific Chemicals
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR423680010
Price (MYR)
560.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
560.00
Add to cart
Specifications
Chemical Name or MaterialEthyl acetate
Melting Point-83.5°C
CAS141-78-6
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.