N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
View all
Glass Bottle
Catalog number ACR423645000
Price (MYR)
560.00
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
560.00
Add to cart
Chemical Identifiers
CAS5810-42-4
IUPAC Nametetrapropylazanium chloride
Molecular FormulaC12H28ClN
InChI KeyFBEVECUEMUUFKM-UHFFFAOYSA-M
SMILES[Cl-].CCC[N+](CCC)(CCC)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration Argentometric>=92 %
Appearance (Form)Crystals or crystalline powder