N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number ACR423640025
Price (MYR)
1,200.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,200.00
Add to cart
Chemical Identifiers
CAS2713-33-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to orange
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)32.0-39.0?C