N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number ACR423640010
Price (MYR)
748.00
Add to cart
Quantity:
1 L
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
748.00
Add to cart
Chemical Identifiers
CAS10039-32-4
IUPAC Namedisodium dodecahydrate hydrogen phosphate
Molecular FormulaH25Na2O16P
InChI KeyDGLRDKLJZLEJCY-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Arsenic (As)=<0.5 ppm
Copper (Cu)=<5 ppm
Magnesium (Mg)=<10 ppm
Nickel (Ni)=<5 ppm