Cupferron, Thermo Scientific Chemicals
Cupferron, Thermo Scientific Chemicals
Cupferron, Thermo Scientific Chemicals
Cupferron, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cupferron, Thermo Scientific Chemicals
CAS: 135-20-6 | C6H9N3O2 | 155.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number ACR423600250
Price (MYR)
539.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
539.00
Add to cart
Chemical Identifiers
CAS67-48-1
IUPAC Name(2-hydroxyethyl)trimethylazanium chloride
Molecular FormulaC5H14ClNO
InChI KeySGMZJAMFUVOLNK-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or needles or adhering crystals
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤2%