Citric acid, 99.6%, Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number ACR423565000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1643-73-8
IUPAC Namechloro[(4-fluorophenyl)methyl]magnesium
Molecular FormulaC7H6ClFMg
InChI KeyPQDGQUPDDGUKLP-UHFFFAOYSA-M
SMILESFC1=CC=C(C[Mg]Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Titration Grignard0.15 to 0.35 M
Appearance (Form)Solution