Citric acid, 99.6%, Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number ACR423560020
Price (MYR)
1,344.00
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (MYR)
1,344.00
Add to cart
Specifications
Chemical Name or MaterialCitric acid
CAS77-92-9
Melting Point153.0°C
Molecular FormulaC6H8O7
Linear FormulaHO2CCH2C(OH)(CO2H)CH2CO2H